
Phosphamidon

Synonym(s):
2-Chloro-N,N-diethyl-3-(dimethylphosphono)crotonic amide
Empirical Formula (Hill Notation):
C10H19ClNO5P
CAS Number:
Molecular Weight:
299.69
Products
Product Category
Brand
Formula Weight
Manufacturer
Product Line
Format
Quality Segment
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing