Skip to Content
Merck
N,N-Diethyl-p-phenylenediamine oxalate salt ≥85% (TLC)

N,N-Diethyl-p-phenylenediamine oxalate salt

N,N-Diethyl-p-phenylenediamine oxalate salt ≥85% (TLC)
Synonym(s):
4-Amino-N,N-diethylaniline oxalate salt
Empirical Formula (Hill Notation):
C10H16N2 · C2H2O4
CAS Number:
Molecular Weight:
254.28

Products

Product Category
Brand
Color
Formula Weight
Melting Point (°C)
Physical Form
Purity
Chemical Composition
Application
Markush Class
Markush Group
Showing 1-1 of 1