
Naloxone solution

Synonym(s):
Naloxone solution
Empirical Formula (Hill Notation):
C19H21NO4
CAS Number:
Molecular Weight:
327.37
Products
Brand
Formula Weight
Gene Alias
Manufacturer
Physical Form
Format
Quality Segment
Application
Package Size
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2