Skip to Content
Merck
Methyl nonanoate analytical standard

Methyl nonanoate

Methyl nonanoate analytical standard
Synonym(s):
Methyl pelargonate, Nonanoic acid methyl ester
Linear Formula:
CH3(CH2)7COOCH3
CAS Number:
Molecular Weight:
172.26
EC Number:

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Purity
Format
Quality Segment
Functional Group
Application
Markush Class
Markush Group
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing