
Mefenamic acid

Synonym(s):
2-[(2,3-Dimethylphenyl)amino]benzoic acid, N-(2,3-Xylyl)anthranilic acid
Empirical Formula (Hill Notation):
C15H15NO2
CAS Number:
Molecular Weight:
241.29
Products
Product Category
Brand
Formula Weight
Gene Alias
Manufacturer
Purity
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Technique
Showing 1-4 of 4