
L-Pyroglutamic acid 2-naphthylamide

Synonym(s):
L-Pyroglutamic acid β-naphthylamide, L-Pyrrolidonyl-β-naphthylamide, N1-(2-Naphthyl)-L-pyroglutamic acid, PYR
Empirical Formula (Hill Notation):
C15H14N2O2
Products
Product Category
Brand
Color
Formula Weight
Physical Form
Purity
Optical Isomer
Amino Acid
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing