
Levomenol

Synonym(s):
(−)-α-Bisabolol, (−)-6-Methyl-2-(4-methyl-3-cyclohexen-1-yl)-5-hepten-2-ol, Levomenol
Empirical Formula (Hill Notation):
C15H26O
CAS Number:
Molecular Weight:
222.37
Products
Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
Package Size
API Family
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing