
Hydrogen chloride solution

Synonym(s):
Hydrochloric acid in methanol
Linear Formula:
HCl · CH3OH
CAS Number:
Molecular Weight:
36.46
Products
Brand
Formula Weight
Quality Segment
Reaction Type
Reagent Type
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing