
Glybenclamide

Synonym(s):
5-Chloro-N-[4-(cyclohexylureidosulfonyl)phenethyl]-2-methoxybenzamide, Glyburide, N-p-[2-(5-Chloro-2-methoxybenzamido)ethyl]benzenesulfonyl-N′-cyclohexylurea
Empirical Formula (Hill Notation):
C23H28ClN3O5S
CAS Number:
Molecular Weight:
494.00
Products
Product Category
Brand
Formula Weight
Gene Alias
Manufacturer
Physical Form
Purity
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-3 of 3