
Furfuryl thioacetate

Synonym(s):
S-Furfuryl thioacetate
Empirical Formula (Hill Notation):
C7H8O2S
CAS Number:
Molecular Weight:
156.20
Products
Product Category
Brand
Biological Source
Boiling Point (°C)
Formula Weight
Purity
Agency
Quality Segment
Application
Markush Class
Markush Group
Organoleptic Class
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing