
Ethambutol Related Compound B

Synonym(s):
(2R,2′R)-2,2-(1,2-Ethanediyldiimino)bis-1-butanol dihydrochloride, levo-Ethambutol hydrochloride
Empirical Formula (Hill Notation):
C10H24N2O2 · 2HCl
CAS Number:
Molecular Weight:
277.23
Products
Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2