
Enalaprilat

Synonym(s):
(2S)-1-[(2S)-2-[[(1S)-1-Carboxy-3-phenyl-propyl]amino]propanoyl]pyrrolidine-2-carboxylic acid, N-[(1S)-1-Carboxy-3-phenylpropyl]-L-alanyl-L-proline
Empirical Formula (Hill Notation):
C18H24N2O5
Molecular Weight:
348.39
Products
Brand
Formula Weight
Gene Alias
Purity
Format
Quality Segment
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing