
Dipyron monohydrate

Synonym(s):
Metamizol, Methapyrone
Empirical Formula (Hill Notation):
C13H16N3NaO4S · H2O
CAS Number:
Molecular Weight:
351.35
Products
Brand
Formula Weight
Gene Alias
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing