![Dextrose Impurity B Pharmaceutical Secondary Standard; Certified Reference Material](/deepweb/assets/sigmaaldrich/product/structures/279/905/0167c99b-56e8-4fae-937a-b620d0cd2d2a/640/0167c99b-56e8-4fae-937a-b620d0cd2d2a.png)
Dextrose Impurity B
![Dextrose Impurity B Pharmaceutical Secondary Standard; Certified Reference Material](/deepweb/assets/sigmaaldrich/product/structures/279/905/0167c99b-56e8-4fae-937a-b620d0cd2d2a/640/0167c99b-56e8-4fae-937a-b620d0cd2d2a.png)
Synonym(s):
Isomaltose, 6-O-α-D-Glucopyranosyl-D-glucose
Empirical Formula (Hill Notation):
C12H22O11
CAS Number:
Molecular Weight:
342.30
Products
Brand
Formula Weight
Physical Form
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing