![Cysteine hydrochloride monohydrate European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/165/000/d03fece8-a3b8-4edd-bdd2-a50b7cae12c1/640/d03fece8-a3b8-4edd-bdd2-a50b7cae12c1.png)
Cysteine hydrochloride monohydrate
![Cysteine hydrochloride monohydrate European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/165/000/d03fece8-a3b8-4edd-bdd2-a50b7cae12c1/640/d03fece8-a3b8-4edd-bdd2-a50b7cae12c1.png)
Synonym(s):
L-Cysteine hydrochloride monohydrate
Linear Formula:
HSCH2CH(NH2)COOH · HCl · H2O
CAS Number:
Molecular Weight:
175.63
Products
Formula Weight
Manufacturer
Format
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing