
Clonazepam

Synonym(s):
5-(2-Chlorophenyl)-7-nitro-3H-1,4-benzodiazepin-2(1H)-one
Empirical Formula (Hill Notation):
C15H10ClN3O3
CAS Number:
Molecular Weight:
315.71
Products
Product Category
Brand
Color
Formula Weight
Gene Alias
Manufacturer
Physical Form
Agency
Format
Research Area
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade