
Clodinafop-propargyl

Synonym(s):
2-Propynyl (R)-2-[4-(5-chloro-3-fluoro-2-pyridinyloxy)phenoxy]propionate
Empirical Formula (Hill Notation):
C17H13ClFNO4
CAS Number:
Molecular Weight:
349.74
Products
Product Category
Brand
Formula Weight
Manufacturer
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2