Calcium ionophore I
Synonym(s):
(−)-(R,R)-N,N′-Bis-[11-(ethoxycarbonyl)undecyl]-N,N′,4,5-tetramethyl-3,6-dioxaoctane-diamide, Diethyl N,N′-[(4R,5R)-4,5-dimethyl-1,8-dioxo-3,6-dioxaoctamethylene]bis(12-methylaminododecanoate), ETH 1001
Empirical Formula (Hill Notation):
C38H72N2O8
Products
Brand
Formula Weight
Product Line
Purity
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing