
Busulfan solution

Empirical Formula (Hill Notation):
C6H14O6S2
CAS Number:
Molecular Weight:
246.30
Products
Brand
Formula Weight
Manufacturer
Physical Form
Format
Quality Segment
Application
Package Size
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing