
Buspirone Related Compound G

Synonym(s):
1,4-Di(pyrimidin-2-yl)piperazine
Empirical Formula (Hill Notation):
C12H14N6
CAS Number:
Molecular Weight:
242.28
Products
Brand
Formula Weight
Manufacturer
Physical Form
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2