
Buprenorphine solution

Synonym(s):
Buprenorphine solution
Empirical Formula (Hill Notation):
C29H41NO4
CAS Number:
Molecular Weight:
467.64
Products
Brand
Formula Weight
Manufacturer
Physical Form
Format
Quality Segment
Application
Package Size
Reference Material Type
Special Grade
Technique
Showing 1-3 of 3
Compare
Product Number
Description
Pricing