![Bupivacaine impurity F European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/296/833/6cc706c0-96b8-435e-8190-6bb73674bcdf/640/6cc706c0-96b8-435e-8190-6bb73674bcdf.png)
Bupivacaine impurity F
![Bupivacaine impurity F European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/296/833/6cc706c0-96b8-435e-8190-6bb73674bcdf/640/6cc706c0-96b8-435e-8190-6bb73674bcdf.png)
Synonym(s):
2,6-Dimethylaniline, 2,6-Xylidine, 2-Amino-1,3-dimethylbenzene, 2-Amino-m-xylene
Linear Formula:
(CH3)2C6H3NH2
CAS Number:
Molecular Weight:
121.18
Products
Boiling Point (°C)
Formula Weight
Manufacturer
Melting Point (°C)
Format
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing