
Bupivacaine impurity E

Synonym(s):
6-(Butylamino)-N-(2,6-dimethylphenyl)hexanamide
Empirical Formula (Hill Notation):
C18H30N2O
CAS Number:
Molecular Weight:
290.44
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing