Skip to Content
Merck
vibrant-m

Bromine water

vibrant-m
Synonym(s):
Bromine solution, Bromide bromate solution
Empirical Formula (Hill Notation):
Br2
CAS Number:
Molecular Weight:
159.81

Products

Brand
Formula Weight
Physical Form
Showing 1-1 of 1
Compare
Product Number
Description
Pricing