Skip to Content
Merck
Brimonidine Related Compound A certified reference material, pharmaceutical secondary standard

Brimonidine Related Compound A

Brimonidine Related Compound A certified reference material, pharmaceutical secondary standard
Synonym(s):
5-Bromoquinoxalin-6-amine
Empirical Formula (Hill Notation):
C8H6BrN3
CAS Number:
Molecular Weight:
224.06

Products

Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade