Skip to Content
Merck
Bis(2-oxo-3-oxazolidinyl)phosphinic chloride ≥97.0% (AT)

Bis(2-oxo-3-oxazolidinyl)phosphinic chloride

Bis(2-oxo-3-oxazolidinyl)phosphinic chloride ≥97.0% (AT)
Synonym(s):
BOP-Cl, Phosphoric acid bis(2-oxooxazolidide) chloride
Empirical Formula (Hill Notation):
C6H8ClN2O5P
CAS Number:
Molecular Weight:
254.56

Products

Product Category
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing