
BIOshell lgG 1000 Å C4 (2.7 μm) HPLC Columns

Products
Product Category
Brand
pH Value
Product Line
Agency
USP Suitability
Analyte Suitability
Separation Technique
ID (mm)
Length (cm)
Quality Segment
Package Size
Particle Size (µm)
Pore Size (Å)
Technique
Showing 1-20 of 23
Page 1 of 2
Compare
Product Number
Description
Pricing
BIOshell lgG 1000 Å C4 (2.7 μm) HPLC Columns, L × I.D. 5 mm × 2.1 mm, HPLC Guard Cartridge, pkg of 3
BIOshell lgG 1000 Å C4 (2.7 μm) HPLC Columns, L × I.D. 5 mm × 4.6 mm, HPLC Guard Cartridge, pkg of 3
Page 1 of 2