Skip to Content
Merck
Apigenin analytical standard

Apigenin

Apigenin analytical standard
Synonym(s):
4′,5,7-Trihydroxyflavone, 5,7-Dihydroxy-2-(4-hydroxyphenyl)-4-benzopyrone, Apigenin
Empirical Formula (Hill Notation):
C15H10O5
CAS Number:
Molecular Weight:
270.24
EC Number:

Products

Product Category
Brand
Color
Formula Weight
Gene Alias
Manufacturer
Melting Point (°C)
Physical Form
Product Line
Purity
Agency
Format
Research Area
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Technique