
Ampicillin

Synonym(s):
D-(−)-α-Aminobenzylpenicillin, Ampicillin
Empirical Formula (Hill Notation):
C16H19N3O4S
CAS Number:
Molecular Weight:
349.40
Products
Product Category
Brand
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Purity
Activity Spectrum
Agency
Format
Quality Segment
Application
Mode of Action
Optical Isomer
Package Size
API Family
Reference Material Type
Special Grade
Technique
Showing 1-5 of 5
Compare
Product Number
Description
Pricing