
Amidosulfuron

Synonym(s):
1-(4,6-Dimethoxypyrimidin-2-yl)-3-(N-mesyl-N-methylsulfamoyl)urea
Empirical Formula (Hill Notation):
C9H15N5O7S2
CAS Number:
Molecular Weight:
369.37
Products
Product Category
Brand
Formula Weight
Manufacturer
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2