![9-Borabicyclo[3.3.1]nonane solution 0.4 M in hexanes](/deepweb/assets/sigmaaldrich/product/structures/180/891/8b64e597-269d-4780-98b6-40889dfd06b9/640/8b64e597-269d-4780-98b6-40889dfd06b9.png)
9-Borabicyclo[3.3.1]nonane solution
![9-Borabicyclo[3.3.1]nonane solution 0.4 M in hexanes](/deepweb/assets/sigmaaldrich/product/structures/180/891/8b64e597-269d-4780-98b6-40889dfd06b9/640/8b64e597-269d-4780-98b6-40889dfd06b9.png)
Synonym(s):
9-BBN
Empirical Formula (Hill Notation):
C8H15B
CAS Number:
Molecular Weight:
122.02
Products
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Quality Segment
Reagent Type
Showing 1-2 of 2
Compare
Product Number
Description
Pricing