
3-epi-25-Hydroxyvitamin D2 solution

Empirical Formula (Hill Notation):
C28H44O2
Products
Product Category
Brand
Color
Formula Weight
Physical Form
Purity
Quality Segment
Application
Markush Class
Markush Group
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing