Skip to Content
Merck
2-Amino-5-hydroxybenzoic acid 98%

2-Amino-5-hydroxybenzoic acid

2-Amino-5-hydroxybenzoic acid 98%
Synonym(s):
5-Hydroxyanthranilic acid
Linear Formula:
HOC6H3(NH2)CO2H
CAS Number:
Molecular Weight:
153.14

Products

Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Application
Markush Class
Markush Group
Amino Acid
Amino Acid Type
Reaction Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing