2,2′-Azobis(2-methylpropionitrile) solution
Synonym(s):
α,α,′-Azoisobutyronitrile solution, AIBN solution
Empirical Formula (Hill Notation):
C8H12N4
CAS Number:
Molecular Weight:
164.21
Products
Brand
Formula Weight
Physical Form
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing