1-Chloro-2-ethynylbenzene
Synonym(s):
(2-Chlorophenyl)acetylene
Linear Formula:
ClC6H4C≡CH
CAS Number:
Molecular Weight:
136.58
Products
Product Category
Brand
Boiling Point (°C)
Formula Weight
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing