
17-α-Hydroxy-3,11-dioxo-androsta-1,4-diene-17-beta-carboxylic acid

Synonym(s):
17α-Hydroxy-3,11-dioxoandrosta-1,4-diene-17β-carboxylic acid
Empirical Formula (Hill Notation):
C20H24O5
CAS Number:
Molecular Weight:
344.40
Products
Brand
Formula Weight
Manufacturer
Product Line
Quality Segment
Application
Special Grade
Showing 1-1 of 1