Skip to Content
Merck
1,3-Bis(tert-butoxycarbonyl)-2-methyl-2-thiopseudourea 98%

1,3-Bis(tert-butoxycarbonyl)-2-methyl-2-thiopseudourea

1,3-Bis(tert-butoxycarbonyl)-2-methyl-2-thiopseudourea 98%
Synonym(s):
N,N′-Di-Boc-S-methylisothiourea, 1,3-Di-Boc-2-methylisothiourea, N,N′-Bis(tert-butoxycarbonyl)-S-methylisothiourea
Linear Formula:
(CH3)3CO2CN=C(SCH3)NHCO2C(CH3)3
CAS Number:
Molecular Weight:
290.38

Products

Formula Weight
Melting Point (°C)
Purity
Application
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing