Z364894
Wheaton media bottles, glass
capacity 1000 mL, PTFE lined cap
Synonym(s):
Wheaton bottles, laboratory bottles, media bottle
About This Item
Recommended Products
material
PTFE cap
borosilicate glass bottle
round bottle
cap size
38-430
Joint
Joints threaded neck (38-430)
description
Autoclavable with cap loose.
sterility
non-sterile
feature
graduations
with cap
packaging
pack of 24 ea
manufacturer/tradename
WHEATON® 219820
H × diam.
231 mm × 103 mm , Cap off size
capacity
1000 mL
Looking for similar products? Visit Product Comparison Guide
General description
- Wheaton media bottles are manufactured from low-alkali borosilicate glass.
- Clear bottles are ideal for storing culture media.
- No-drip lip for clean pouring.
- Sloping shoulder for easier cleaning.
- Marking patch for identification.
- Graduations show approximate volume.
- Includes an autoclavable cap for sample protection.
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service