Key Documents
OH30073972
Ohaus® Pen pH Meter
Model ST10C-A
About This Item
UNSPSC Code:
41115603
packaging
pack of 1 ea
manufacturer/tradename
Ohaus® 30073972
conductivity measuring range
0.00-199.9 μS/cm
battery power
4 × 1.5 V, 200 hr life
General description
With durable IP67 waterproof ABS housing, protective sensor cap, and an automatic shutdown feature that preserves battery life, OHAUS pen meters can endure consistent use in rough, wet environments.
Application
pH, oxidation-reduction potential (ORP), conductivity, dissolved oxygen (DO), salinity, total dissolved solids (TDS), temperature measurement
Features and Benefits
Easily replaceable electrodes, automatic shut off feature
Legal Information
Ohaus is a registered trademark of Ohaus Corp.