MAK481
Ethanol Assay Kit
sufficient for 100 colorimetric tests
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12161503
NACRES:
NA.84
Recommended Products
Quality Level
application(s)
pharmaceutical
detection method
colorimetric
relevant disease(s)
gastrointestinal diseases
shipped in
wet ice
storage temp.
−20°C
General description
Alcoholic drinks are among the daily consumed beverages. Studies have shown that heavy alcohol consumption may lead to various forms of liver diseases and to increased mortality rates.
Features and Benefits
- The linear detection range of the kit is 0.0008% (v/v) (140 µM or 8 ppm) to 0.1% (v/v).
- Compatible with high-throughput handling systems.
Suitability
The kit is suitable for the quantitative determination of ethanol in serum, plasma, urine, and saliva samples, as well as studying the effects of drugs on alcohol metabolism.
Principle
The Ethanol Assay Kit is based on the alcohol dehydrogenase-catalyzed oxidation of ethanol, in which the formed NADH is coupled to the formazan (MTT) chromogen. The intensity of the product color, measured at 565 nm, is proportionate to the ethanol concentration in the sample.
related product
Product No.
Description
Pricing
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Met. Corr. 1
Storage Class Code
8A - Combustible corrosive hazardous materials
WGK
WGK 2
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service