NIST1155A
Stainless Steel
NIST® SRM® 1155a, Cr 18 - Ni 12 - Mo 2 (AISI 316)
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
grade
certified reference material
Quality Level
packaging
pkg of disk
manufacturer/tradename
NIST®
format
matrix material
Looking for similar products? Visit Product Comparison Guide
General description
Each unit of SRM 1155a is a solid disk measuring 3.2 cm in diameter and 1.9 cm in thickness.
SRM 1155A_cert
SRM 1155A_SDS
SRM 1155A_cert
SRM 1155A_SDS
Application
- HPLC-UV and GC-MS Methods for Determination of Chlorambucil and Valproic Acid in Plasma for Further Exploring a New Combined Therapy of Chronic Lymphocytic Leukemia.: This study employed the LiChrospher® 100 RP-18 endcapped (10 µm) LICHROCART™ 250-4 column for the high-performance liquid chromatography analysis of chlorambucil and valproic acid in plasma. The method facilitated efficient separation and detection, critical for evaluating combined chemotherapy protocols (Lipska K et al., 2021).
- Application of chemometric approach for development and validation of high performance liquid chromatography method for estimation of ropinirole hydrochloride.: This research utilized the LiChrospher® 100 RP-18 endcapped column to develop and validate a robust HPLC method for the quantification of ropinirole hydrochloride. The study demonstrated the column′s capability in achieving high sensitivity and precision in pharmaceutical analysis (Fatima S et al., 2019).
- Determination of retinol and retinyl esters in human plasma by high-performance liquid chromatography with automated column switching and ultraviolet detection.: The LiChrospher® 100 RP-18 endcapped column was applied in the analysis of retinol and its esters in human plasma. This study showcased the column′s efficiency in separating closely related compounds, essential for accurate biochemical assays (Hartmann S et al., 2001).
- Rapid, highly sensitive method for the determination of morphine and its metabolites in body fluids by liquid chromatography-mass spectrometry.: This investigation leveraged the LiChrospher® 100 RP-18 endcapped column to develop a sensitive LC-MS method for morphine and its metabolites. The column′s performance ensured reliable detection in forensic and clinical toxicology (Schänzle G et al., 1999).
- Determination of 13-cis-3-hydroxyretinoic acid, all-trans-3-hydroxyretinoic acid and their 4-oxo metabolites in human and animal plasma by high-performance liquid chromatography with automated column switching and UV detection.: This study used the LiChrospher® 100 RP-18 endcapped column for the simultaneous analysis of retinoic acid metabolites, highlighting its application in pharmacokinetics and metabolism research (Wyss R et al., 1998).
- Determination of endogenous levels of 13-cis-retinoic acid (isotretinoin), all-trans-retinoic acid (tretinoin) and their 4-oxo metabolites in human and animal plasma by high-performance liquid chromatography with automated column switching and ultraviolet detection.: The research focused on using the LiChrospher® 100 RP-18 endcapped column for analyzing endogenous retinoic acids in plasma, demonstrating the column′s utility in clinical biochemistry and toxicology studies (Wyss R et al., 1997).
Features and Benefits
- Accompanied with certificate documenting NIST-certified and reference mass fraction values, and coverage factor(k) for elements in stainless steel matrix of SRM1155A.
- Information values for elements are reported as mass fractions.
- The certified values are metrologically traceable to the derived SI unit of mass, expressed in %.
- The NIST certificate is provided with expiration certificate, storage, handling,use, and maintenance instructions.
Other Notes
Certified for the analytes listed below.
Carbon (C), Cobalt (Co), Chromium (Cr), Copper (Cu), Iron (Fe), Manganese (Mn), Molybdenum (Mo), Niobium (Nb), Nickel (Ni), Phosphorus (P), Silicon (Si), Titanium (Ti), Vanadium (V), Tungsten (W).
Reference/Informational value is provided for the following analytes.
Arsenic (As), Nitrogen (N), Sulfur (S), Tin (Sn), Aluminum (Al), Boron (B), Bismuth (Bi), Lead (Pb), Oxygen (O)
See certificate for values and more details at nist.gov/SRM.
Carbon (C), Cobalt (Co), Chromium (Cr), Copper (Cu), Iron (Fe), Manganese (Mn), Molybdenum (Mo), Niobium (Nb), Nickel (Ni), Phosphorus (P), Silicon (Si), Titanium (Ti), Vanadium (V), Tungsten (W).
Reference/Informational value is provided for the following analytes.
Arsenic (As), Nitrogen (N), Sulfur (S), Tin (Sn), Aluminum (Al), Boron (B), Bismuth (Bi), Lead (Pb), Oxygen (O)
See certificate for values and more details at nist.gov/SRM.
Legal Information
LICHROCART is a trademark of Merck KGaA, Darmstadt, Germany
LiChrospher is a registered trademark of Merck KGaA, Darmstadt, Germany
NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology
related product
Product No.
Description
Pricing
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service