Skip to Content
Merck
All Photos(2)

Key Documents

I0765

Sigma-Aldrich

Immersion oil

viscosity 150 cSt (lit.)

Synonym(s):

immersion medium for light microscopy

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12171500
NACRES:
NA.47

biological source

synthetic

Quality Level

color

yellow

viscosity

150 cSt(lit.)

density

0.9 g/cm3 at 23 °C

application(s)

hematology
histology

storage temp.

room temp

General description

Immersion Oil has a defined refractive index of about 1.516. It is important that the refractive index (nD) is about 1.5, similar to that of glass. This enables a homogeneous oil immersion to be achieved. Immersion media are used in conjunction with immersion objectives and are located between the surface of the specimen and the lens of the microscope. Immersion oils practically eliminate light beam deflection so that the effectiveness of the lens is considerably increased.

Application

Immersion oil has been used in the following applications:
  1. in epifluorescent microscopy and cell identification
  2. in the assembly and filling of the microinjecting system
  3. in microscopy to reliably visualize dendritic spines

Biochem/physiol Actions

Immersion oil has a larger impact on 3D resolution. Specifically, matching the immersion oil refractive index to glass, reliably enhances the ability to visualize dendritic spines.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

159.8 °F

Flash Point(C)

71 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Impact of immersion oils and mounting media on the confocal imaging of dendritic spines.
Peterson BM, et al.
Journal of Neuroscience Methods, 242, 106-111 (2015)
A simple pressure microinjecting system for delivery of small substance volumes to the brain: application to the developmental study of thalamo-cortical projections in foetal and neonatal rats.
Sbriccoli A, et al.
Brain Res. Protoc., 3(3), 257-263 (1999)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service