Skip to Content
Merck
All Photos(1)

Key Documents

AB3831-I

Sigma-Aldrich

Anti-phospho Cofilin-1 Antibody (Ser3)

from rabbit, purified by affinity chromatography

Synonym(s):

Cofilin-1, Ser3-phosphorylated, 18 kDa phosphoprotein, Ser3-phosphorylated, Cofilin, non-muscle isoform, Ser3-phosphorylated

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

species reactivity (predicted by homology)

bovine (based on 100% sequence homology), porcine (based on 100% sequence homology), mouse (based on 100% sequence homology), rat (based on 100% sequence homology)

technique(s)

inhibition assay: suitable (peptide)
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

phosphorylation (pSer3)

Gene Information

human ... CFL1(1072)

General description

Cofilin-1 (UniProt P23528; also known as 18 kDa phosphoprotein, Cofilin non-muscle isoform, p18) is encoded by the CFL1 (also known as CFL) gene (Gene ID 1072) in human. Cofilin-1 (non-muscle cofilin), cofilin-2 (muscle cofilin), and actin-depolymerizing factor (ADF) play an essential role in regulating actin filament dynamics and reorganization by stimulating the severance and depolymerization of actin filaments. The three cofilin family members share 70% to 81% sequence identity, exhibit similar biological functions, and display distinct but overlapping expression patterns. Cofilins and ADF bind actin monomers and filaments (G-actin and F-actin) and increase the number of actin monomers and filament fragments, facilitating filament turnover and treadmilling at key locations in migrating cells. Cofilins and ADF are inactivated by phosphorylation at Ser3 by LIM-kinases (LIMKs) and testicular protein kinases (TESKs) and are reactivated by dephosphorylation by the slingshot (SSH) family of protein phosphatases and chronophin. Cfl1-knockout results in mouse embryonic lethality at E11.5 12.5 due to aberrant neural tube closure and defective neural crest cell migration, while Adf-deficient mice are viable with normal brain appearance, but suffer from corneal defects that cause blindness in adult mice.

Specificity

Target band detection by this polyclonal antibody was prevented by preblocking with the immunogen phosphopeptide, but not the corresponding non-phosphopeptide.

Immunogen

KLH-conjugated linear peptide corresponding to the N-termnal sequence of human cofilin-1 with phosphorylated Ser3.

Application

Peptide Inhibition Assay: Target band detection in 10 µg of HeLa cell lysate was prevented by preblocking a representative lot with the immunogen phosphopeptide, but not the corresponding non-phosphopeptide.
Research Category
Cell Structure
This rabbit polyclonal Anti-phospho Cofilin-1 (Ser3), Cat. No. AB3831-I is validated for use in Peptide Inhibition Assay and Western Blotting for the detection of Cofilin-1 Ser3 phosphorylation.

Quality

Evaluated by Western Blotting in HeLa cell lysate.

Western Blotting Analysis: A 1:200 dilution of this antibody detected Ser3-phosphorylatd cofilin-1 in 10 µg of HeLa cell lysate.

Target description

~19 kDa observed. 18.39/18.37/184.3/18.39/18.40 kDa (bovine/human/mouse/porcine/rat) calculated (Met1 removed). Uncharacterized bands may be observed in some lysate(s).

Physical form

Affinity purified.
Purified rabbit polyclonal antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service