Skip to Content
Merck
All Photos(1)

Key Documents

1.01515

Supelco

Silver diethyldithiocarbamate

for analysis EMSURE®,Reag. Ph Eur

Synonym(s):

Silver diethyldithiocarbamate, Diethyldithiocarbamic acid silver salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H10AgNS2
CAS Number:
Molecular Weight:
256.14
MDL number:
UNSPSC Code:
12352107
EC Index Number:
216-003-7
NACRES:
NA.05

Agency

reag. Ph. Eur.

Quality Level

product line

EMSURE®

Assay

≥99 % ((DSC (differential scanning calorimetry)))

form

solid

pH

5.0-6.5 ( in H2O, suspension)

mp

178-183 °C

bulk density

340 kg/m3

storage temp.

2-8°C

SMILES string

CCN(CC)C(=S)[S-].[Ag+]

InChI

1S/C5H11NS2.Ag/c1-3-6(4-2)5(7)8;/h3-4H2,1-2H3,(H,7,8);/q;+1/p-1

InChI key

NSVHDIYWJVLAGH-UHFFFAOYSA-M

Related Categories

Application


  • Arsenite tolerance and removal potential of the indigenous halophilic bacterium, Halomonas elongata SEK2.: This study explores the application of silver diethyldithiocarbamate in bioremediation, specifically its role in enhancing arsenite removal by Halomonas elongata SEK2. This research provides insights into the potential of using complexed silver compounds in environmental cleanup efforts, particularly in saline environments where traditional methods may fail. The findings could significantly impact wastewater treatment and environmental sustainability practices (Tavoosi et al., 2024).

  • The development and use of an innovative laboratory method for measuring arsenic in drinking water from western Bangladesh.: This paper details the use of silver diethyldithiocarbamate in a novel analytical method to measure trace levels of arsenic in drinking water. This development not only enhances the accuracy and sensitivity of arsenic detection but also demonstrates the critical role of silver diethyldithiocarbamate in public health monitoring, offering a significant improvement over traditional testing methods (Frisbie et al., 2005).

  • Methods of analysis for toxic elements in food products. 2. Review of USSR standards on determinations of heavy metals and arsenic.: This review discusses various methods, including the use of silver diethyldithiocarbamate, for the analysis of toxic elements in food products. The article highlights the importance of stringent quality control in food safety and the role of advanced chemical reagents in ensuring the reliability of detection methods in the food industry (Skurikhin, 1989).

Analysis Note

Purity (DSC (differential scanning calorimetry)): ≥ 99 %
Identity (IR-spectrum): passes test
Appearance of solution (in pyridine): passes test
pH-value (aqueous suspension): 5.0 - 6.5
Melting range (DSC (differential scanning calorimetry)): 178 - 183 °C
Spec. Absorptivity A 1%/1cm (of the As-complex; 525 nm; 0.00033 %; 1 cm; pyridine): ≥ 1500
Nitrate (NO₃): ≤ 0.01 %
Loss on Drying (105 °C): ≤ 0.5 %
Sensitivity (for determination of arsenic): passes test

Legal Information

EMSURE is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 2

1 of 2

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service