Skip to Content
Merck
All Photos(1)

Key Documents

CLS7082516

Pyrex® VISTA culture tube, screw-cap

diam. × L 16 mm × 100 mm

Synonym(s):

culture tubes, test tubes, test tubes glass, Corning® 70825

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121706
NACRES:
NB.43

material

borosilicate glass
cap (Rubber liner)
round bottom tube

sterility

non-sterile

packaging

case of 200

manufacturer/tradename

Corning 70825-16

diam. × L

16 mm × 100 mm

color

clear

Looking for similar products? Visit Product Comparison Guide

General description

These Pyrex Vista culture tubes are supplied with deep-form phenolic caps to facilitate handling and sealing after autoclaving. The blue enamel marking spot gives an excellent surface for pencil notations. Pencil marks are easily erased.

Legal Information

Corning is a registered trademark of Corning, Inc.
PYREX is a registered trademark of Corning, Inc.
VISTA is a trademark of Corning, Inc.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service