Skip to Content
Merck
All Photos(1)

Key Documents

95395

Sigma-Aldrich

Tungstosilicic acid hydrate

purum p.a.

Synonym(s):

Silicotungstic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H4[Si(W3O10)4] · xH2O
CAS Number:
Molecular Weight:
2878.17 (anhydrous basis)
MDL number:
UNSPSC Code:
12352301
PubChem Substance ID:
NACRES:
NA.22

grade

purum p.a.

form

powder or crystals

loss

≤20% loss on ignition

solubility

water: 0.1 g/mL, clear, colorless

anion traces

chloride (Cl-): ≤200 mg/kg
sulfate (SO42-): ≤100 mg/kg

cation traces

Ca: ≤200 mg/kg
Cd: ≤50 mg/kg
Co: ≤50 mg/kg
Cu: ≤50 mg/kg
Fe: ≤50 mg/kg
K: ≤200 mg/kg
Na: ≤3000 mg/kg
Ni: ≤50 mg/kg
Pb: ≤50 mg/kg
Zn: ≤50 mg/kg

SMILES string

O.O[Si](O)(O)O.O=[W]1(=O)O[W](=O)(=O)O[W](=O)(=O)O1.O=[W]2(=O)O[W](=O)(=O)O[W](=O)(=O)O2.O=[W]3(=O)O[W](=O)(=O)O[W](=O)(=O)O3.O=[W]4(=O)O[W](=O)(=O)O[W](=O)(=O)O4

InChI

1S/H4O4Si.H2O.36O.12W/c1-5(2,3)4;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;/h1-4H;1H2;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;

InChI key

ACTPFSYIGLFGLY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Reactant for:
  • Preparation of silicotungstate butylmethylimidazolium bromide ionic liquid hybrid material
  • Synthesis of SnII-substituted Keggin-type polyoxometalates as photocatalysts
  • Preparation of silicotungstic acid/organically modified silane proton-conducting membranes
  • Preparation of charge transfer compounds with Keggin anions
  • Preparation of catalyst for catalytic liquid-phase oxidation of xylene

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service