Skip to Content
Merck
All Photos(1)

Key Documents

920363

Sigma-Aldrich

Lithium hexafluoroantimonate(V)

98%

Synonym(s):

Lithium hexafluoroantimonate, Antimony lithium fluoride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
LiSbF6
CAS Number:
Molecular Weight:
242.69
MDL number:
UNSPSC Code:
12352302
NACRES:
NA.23

Quality Level

Assay

98%

SMILES string

[Li+].F[Sb-](F)(F)(F)(F)F

InChI

1S/6FH.Li.Sb/h6*1H;;/q;;;;;;+1;+5/p-6

InChI key

YVBBFYDVCPCHHG-UHFFFAOYSA-H

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

LiSbF6 can be used for creating crystalline polymer electrolytes.Amorphous aliphatic polycarbonate/lithium hexafluoroantimonate solid state electrolytes exhibit good mechanical properties and very transparent appearance giving broad applications in displays .

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jianjun Zhang et al.
Small (Weinheim an der Bergstrasse, Germany), 14(36), e1800821-e1800821 (2018-08-04)
Conventional liquid electrolytes based lithium-ion batteries (LIBs) might suffer from serious safety hazards. Solid-state polymer electrolytes (SPEs) are very promising candidate with high security for advanced LIBs. However, the quintessential frailties of pristine polyethylene oxide/lithium salts SPEs are poor ionic
Z Gadjourova et al.
Nature, 412(6846), 520-523 (2001-08-03)
Polymer electrolytes are the subject of intensive study, in part because of their potential use as the electrolyte in all-solid-state rechargeable lithium batteries. These materials are formed by dissolving a salt (for example LiI) in a solid host polymer such

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service