Skip to Content
Merck
All Photos(1)

Key Documents

366501

Sigma-Aldrich

Ammonium-15N,d4 chloride

98 atom % 15N, 99 atom % D

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
15ND4Cl
CAS Number:
Molecular Weight:
58.51
MDL number:
UNSPSC Code:
12142200
PubChem Substance ID:
NACRES:
NA.12

isotopic purity

98 atom % 15N
99 atom % D

form

powder

technique(s)

protein expression: suitable

mp

340 °C (subl.) (lit.)

mass shift

M+5

SMILES string

[Cl-].[2H][15N+]([2H])([2H])[2H]

InChI

1S/ClH.H3N/h1H;1H3/i;1+1/hD4

InChI key

NLXLAEXVIDQMFP-QJNGTKNVSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service