Merck
Paraneoplastic neuromyelitis optica spectrum disorder associated with metastatic carcinoid expressing aquaporin-4.
JAMA neurology (2014-04-16)
Michelle Figueroa, Yong Guo, Alexandros Tselis, Sean J Pittock, Vanda A Lennon, Claudia F Lucchinetti, Robert P Lisak
RÉSUMÉ
Reports of neuromyelitis optica spectrum disorder (NMOSD) occurring in the setting of neoplasia suggest that aquaporin-4 autoimmunity may in some cases have a paraneoplastic basis. In this case report, we describe a patient with NMOSD whose test results were seropositive for aquaporin-4 IgG and who had a hepatic metastasis from a small-bowel neuroendocrine tumor. The tumor cells expressed aquaporin-4 immunoreactivity. She presented to the Neurology Department at Wayne State University with bilateral leg weakness, ascending paresthesias, and decreased sensation. This case extends the context of NMOSD as a paraneoplastic disorder.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
Anti-Water Channel Aquaporin 4 antibody produced in rabbit, lyophilized powder, affinity isolated antibody