Merck
  • Evaluation of short interval cortical inhibition and intracortical facilitation from the dorsolateral prefrontal cortex in patients with schizophrenia.
Evaluation of short interval cortical inhibition and intracortical facilitation from the dorsolateral prefrontal cortex in patients with schizophrenia.
Scientific reports (2017-12-08)
Yoshihiro Noda, Mera S Barr, Reza Zomorrodi, Robin F H Cash, Faranak Farzan, Tarek K Rajji, Robert Chen, Zafiris J Daskalakis, Daniel M Blumberger
RÉSUMÉ
GABAergic and glutamatergic dysfunction in the dorsolateral prefrontal cortex (DLPFC) are thought to be the core pathophysiological mechanisms of schizophrenia. Recently, we have established a method to index these functions from the DLPFC using the paired transcranial magnetic stimulation (TMS) paradigms of short interval intracortical inhibition (SICI) and facilitation (ICF) combined with electroencephalography (EEG). In this study, we aimed to evaluate neurophysiological indicators related to GABA
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
Ethyltriphenylphosphonium bromide, 99%
Sigma-Aldrich
1,3-Propylene sulfite, 99%